Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLN3 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549382

Catalog No. PA549382


Add to cart

Description

Description

Predicted to react with monkey based on sequence homology.

This gene encodes a protein that is involved in lysosomal function. Mutations in this, as well as other neuronal ceroid-lipofuscinosis (CLN) genes, cause neurodegenerative diseases commonly known as Batten disease or collectively known as neuronal ceroid lipofuscinoses (NCLs). Many alternatively spliced transcript variants have been found for this gene. [provided by RefSeq].
Specifications

Specifications

CLN3
Polyclonal
PBS with 0.09% sodium azide
Q13286, Q61124
Battenin, Batten disease protein, Protein CLN3, CLN3, BTS
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
1201, 12752
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
CLN3
Liquid
CLN3
KLH conjugated synthetic peptide between 250-284 amino acids from the Central region of human CLN3.
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit