Learn More
Aviva Systems Biology Corp CLN8 Antibody - N-terminal region (ARP49408_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP49408_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CLN8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CLN8. Alternate names: C8orf61, EPMR, FLJ39417ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human,
Catalog No. 50-014-60077