Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

CLN8 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP19133420UL

 View more versions of this product

Catalog No. NBP19133420



Description

Description

CLN8 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

CLN8
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
CLN8
Synthetic peptide directed towards the N terminal of human CLN8. Peptide sequence VFGVQSTAAGLWALLGDPVLHADKARGQQNWCWFHITTATGFFCFENVAV.
Immunogen affinity purified
RUO
Primary
2055
Western Blot
Western Blot 1:1000
NP_061764
C8orf61, ceroid-lipofuscinosis, neuronal 8 (epilepsy, progressive with mental retardation), EPMR
Rabbit
IgG
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit