Learn More
CLOCK Mouse anti-Human, Clone: OTI1G3, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF804782
Catalog No. 50-166-7133
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a protein that belongs to the basic helix-loop-helix (bHLH) family of transcription factors. Polymorphisms within the encoded protein have been associated with circadian rhythm sleep disorders. A similar protein in mice is a circadian regulator that acts as a transcription factor and forms a heterodimer with aryl hydrocarbon receptor nuclear translocator-like to activate transcription of mouse period 1.
Specifications
Specifications
CLOCK
Monoclonal
Unconjugated
CLOCK
Lyophilized
CLOCK
Human recombit protein fragment corresponding to amino acids 1-285 of human CLOCK produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
OTI1G3
PBS with 8% trehalose and no preservative; pH 7.3
O15516
KAT13D, bHLHe8
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
9575