Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLOCK Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 180941AP

Catalog No. 50-173-1981


Add to cart

Description

Description

This gene encodes a protein that belongs to the basic helix-loop-helix (bHLH) family of transcription factors. Polymorphisms within the encoded protein have been associated with circadian rhythm sleep disorders. A similar protein in mice is a circadian regulator that acts as a transcription factor and forms a heterodimer with aryl hydrocarbon receptor nuclear translocator-like to activate transcription of mouse period 1.
Specifications

Specifications

CLOCK
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O15516
bHLHe8, CLOCK, clock homolog (mouse), hCLOCK, KAT13D, KIAA0334
Rabbit
IgG
150 μL
-20°C
Primary
9575
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
CLOCK
Liquid
CLOCK
CLOCK Fusion Protein Ag12826
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit