Learn More
Clofarabine, Tocris Bioscience™
Deoxycytidine kinase (dCK) substrate
Manufacturer:  Tocris Bioscience 2600/25
 View more versions of this product
Catalog No. 26-002-5
Add to cart
This item is not returnable. View return policy
Description
Description
Deoxycytidine kinase (dCK) substrate. Phosphorylated to form clofarabine triphosphate, which competes with dATP for DNA polymerase-α and -ε and potently inhibits ribonucleotide reductase (IC50 = 65 nM). Induces apoptosis by directly altering mitochondrial transmembrane potential. Demonstrates growth inhibition and cytotoxic activity in a variety of leukemias and solid tumors.
Specifications
Specifications
2-Chloro-9-(2-deoxy-2-fluoro-β-D-arabinofuranosyl)-9H-purin-6-amine
>99%
Observed MW: 303.68
123318-82-1
C10H11ClFN5O3
25mg
Safety and Handling
Safety and Handling
Recommended Storage : Desiccate at 4°C