Learn More
Selleck Chemical LLCSupplier Diversity Partner Clofarabine 10mg 123318-82-1  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S121810MG
Clofarabine inhibits the enzymatic activities of ribonucleotide reductase (RNR) (IC50 = 65 nM) and DNA polymerase. Clofarabine induces autophagy and apoptosis.
Catalog No. 50-673-8