Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Clofarabine
Deoxycytidine kinase (dCK) substrate
$165.88 - $620.60
Specifications
Chemical Name or Material | 2-Chloro-9-(2-deoxy-2-fluoro-β-D-arabinofuranosyl)-9H-purin-6-amine |
---|---|
CAS | 123318-82-1 |
Assay Percent Range | >99% |
Molecular Formula | C10H11ClFN5O3 |
Formula Weight | Observed MW: 303.68 |
Description
Deoxycytidine kinase (dCK) substrate. Phosphorylated to form clofarabine triphosphate, which competes with dATP for DNA polymerase-α and -ε and potently inhibits ribonucleotide reductase (IC50 = 65 nM). Induces apoptosis by directly altering mitochondrial transmembrane potential. Demonstrates growth inhibition and cytotoxic activity in a variety of leukemias and solid tumors.Specifications
2-Chloro-9-(2-deoxy-2-fluoro-β-D-arabinofuranosyl)-9H-purin-6-amine | |
>99% | |
Observed MW: 303.68 |
123318-82-1 | |
C10H11ClFN5O3 |
Safety and Handling
Recommended Storage : Store at 4°C
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title