Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clofibric acid, MP Biomedicals™

Catalog No. p-4679710
Click to view available options
Quantity:
10 g
100 g

Chemical Identifiers

CAS 882-09-7
Molecular Formula C10H11ClO3
Molecular Weight (g/mol) 214.645
InChI Key TXCGAZHTZHNUAI-UHFFFAOYSA-N
Synonym clofibric acid, 2-4-chlorophenoxy-2-methylpropanoic acid, chlorofibrinic acid, clofibrinic acid, chlorfibrinic acid, clofibrin, chlorophibrinic acid, pcib, 2-4-chlorophenoxy-2-methylpropionic acid, clofibrate free acid
PubChem CID 2797
ChEBI CHEBI:34648
IUPAC Name 2-(4-chlorophenoxy)-2-methylpropanoic acid
SMILES CC(C)(C(=O)O)OC1=CC=C(C=C1)Cl

Specifications

Quantity 10 g
Formula Weight 214.6
Physical Form Solid
Chemical Name or Material Clofibric acid
Recommended Storage Store at 2° to 8°C.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.