Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clomipramine hydrochloride, Tocris Bioscience™

Click to view available options
Quantity:
500 mg
This item is not returnable. View return policy
17321-77-6
C19H24Cl2N2
351.315
WIMWMKZEIBHDTH-UHFFFAOYSA-N
clomipramine hydrochloride, anafranil, clomipramine hcl, 3-3-chloro-10,11-dihydro-5h-dibenzo b,f azepin-5-yl-n,n-dimethylpropan-1-amine hydrochloride, anaphranil, 3-chloroimipramine hydrochloride, chloroimipramine monohydrochloride, chlorimipramine hydrochloride, unii-2lxw0l6gwj
68539
CHEBI:3755
3-(2-chloro-5,6-dihydrobenzo[b][1]benzazepin-11-yl)-N,N-dimethylpropan-1-amine;hydrochloride
CN(C)CCCN1C2=CC=CC=C2CCC3=C1C=C(C=C3)Cl.Cl
This item is not returnable. View return policy
17321-77-6
C19H24Cl2N2
351.315
WIMWMKZEIBHDTH-UHFFFAOYSA-N
clomipramine hydrochloride, anafranil, clomipramine hcl, 3-3-chloro-10,11-dihydro-5h-dibenzo b,f azepin-5-yl-n,n-dimethylpropan-1-amine hydrochloride, anaphranil, 3-chloroimipramine hydrochloride, chloroimipramine monohydrochloride, chlorimipramine hydrochloride, unii-2lxw0l6gwj
68539
CHEBI:3755
3-(2-chloro-5,6-dihydrobenzo[b][1]benzazepin-11-yl)-N,N-dimethylpropan-1-amine;hydrochloride
CN(C)CCCN1C2=CC=CC=C2CCC3=C1C=C(C=C3)Cl.Cl

5-HT reuptake inhibitor

Potent, selective 5-HT uptake blocker. Antidepressant. Binds to the human 5-HT transporter with a Ki of 0.05 nmol/l.
Quantity 500 mg
Formula Weight 351.32
Percent Purity >98%
Chemical Name or Material Clomipramine hydrochloride

Recommended Storage : Store at Room Temperature

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit