Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clomipramine Hydrochloride, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog Number 18601752
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
17321-77-6
C19H24Cl2N2
351.32
WIMWMKZEIBHDTH-UHFFFAOYSA-N
hydrogen (3-{5-chloro-2-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-2-yl}propyl)dimethylamine chloride
[H+].[Cl-].CN(C)CCCN1C2=CC=CC=C2CCC2=CC=C(Cl)C=C12
17321-77-6
C19H24Cl2N2
351.32
WIMWMKZEIBHDTH-UHFFFAOYSA-N
hydrogen (3-{5-chloro-2-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-2-yl}propyl)dimethylamine chloride
[H+].[Cl-].CN(C)CCCN1C2=CC=CC=C2CCC2=CC=C(Cl)C=C12

C1376, 17321-77-6, C19H23ClN2•HCl

Spectrum™ Chemical Clomipramine Hydrochloride, USP has a trade name of Anafranil and has several uses in medicine including the treatment of panic disorder, major depressive disorder, and chronic pain. The USP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 1%
Packaging Poly Pail
Quantity 1 kg
Percent Purity 98-102%
Grade USP
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit