Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clomipramine Hydrochloride, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
17321-77-6
C19H24Cl2N2
351.32
WIMWMKZEIBHDTH-UHFFFAOYSA-N
hydrogen (3-{5-chloro-2-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-2-yl}propyl)dimethylamine chloride
[H+].[Cl-].CN(C)CCCN1C2=CC=CC=C2CCC2=CC=C(Cl)C=C12
17321-77-6
C19H24Cl2N2
351.32
WIMWMKZEIBHDTH-UHFFFAOYSA-N
hydrogen (3-{5-chloro-2-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-2-yl}propyl)dimethylamine chloride
[H+].[Cl-].CN(C)CCCN1C2=CC=CC=C2CCC2=CC=C(Cl)C=C12

C1376, 17321-77-6, C19H23ClN2•HCl

Spectrum™ Clomipramine Hydrochloride, USP has a trade name of Anafranil and has several uses in medicine including the treatment of panic disorder, major depressive disorder, and chronic pain. The USP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Quantity 5 kg
Loss on Drying 1%
Ignition Residue 0.1%
Packaging Poly Pail
CAS Max % 100%
Percent Purity 98-102%