Learn More
Clonidine hydrochloride, 98+%, Thermo Scientific Chemicals
Description
Clonidine hydrochloride tablets are indicated in the treatment of hypertension. It has found new uses, including treatment of some types of neuropathic pain, opioid detoxification, sleep hyperhidrosis, anaesthetic use, and off-label, to counter the side effects of stimulant medications such as methylphenidate or amphetamine. It is becoming a more accepted treatment for insomnia, as well as for relief of menopausal symptoms. Clonidine(4205-91-8) is increasingly used in conjunction with stimulants to treat attention-deficit hyperactivity disorder (ADHD).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Soluble in water (50 mg/ml), DMSO (75 mM), methanol, chloroform (slightly), and dehydrated alcohol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Color | White |
Quantity | 1 g |
UN Number | UN2811 |
Beilstein | 4163525 |
Merck Index | 14,2390 |
Solubility Information | Soluble in water (50mg/ml),DMSO (75 mM),methanol,chloroform (slightly),and dehydrated alcohol. |
Formula Weight | 266.55 |
Percent Purity | ≥98% |
Physical Form | Powder |
Chemical Name or Material | Clonidine hydrochloride |
Safety and Handling
Hazard Category | H301-H330 |
Hazard Statement | GHS H Statement H301-H330 Toxic if swallowed. Fatal if inhaled. |
Precautionary Statement | P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P330-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S. |
EINECSNumber | 224-121-5 |
RTECSNumber | NJ2490000 |
TSCA | No |
Recommended Storage | Keep cold |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.