Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clostridium Botulinum E Toxoid Mouse anti-Bacteria, Alexa Fluor™ 532, Clone: 8B9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP223515AF532

Catalog No. NP2231A32


Add to cart

Description

Description

Clostridium Botulinum E Toxoid Monoclonal antibody specifically detects Clostridium Botulinum E Toxoid in Bacteria samples. It is validated for ELISA.
Specifications

Specifications

Clostridium Botulinum E Toxoid
8B9
ELISA
BoNT/E, Bontoxilysin e, Botulinum neurotoxin type e non toxic component, Botulinum neurotoxin type e precursor, Botulism E toxoid, Ent120
Clostridium botulinum type E toxoid adsorbed onto aluminum hydroxide gel
0.1 mL
Primary
Clone 8B9 is specific for botulinum toxoid. No cross-reactivity to type A and type F toxoids.
ELISA
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Bacteria
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit