Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clostridium Difficile Toxin A Mouse anti-Bacteria, Clone: 253/422, Novus Biologicals™

Mouse Monoclonal Antibody

Specifications

Antigen Clostridium Difficile Toxin A
Clone 253/422
Concentration 0.2 mg/ml
Dilution ELISA 1-5 ug/ml, Immunocytochemistry/Immunofluorescence 1-2 ug/ml
Applications ELISA, Immunofluorescence
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122540
View Documents
Novus Biologicals
NBP30753320UG
20 μg
Each for $138.37
N/A
Due to product restrictions, please to purchase or view availability for this product.
NB122539
View Documents
Novus Biologicals
NBP30753310UG
100 μg
Each for $352.89
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description

Description

Clostridium Difficile Toxin A Monoclonal antibody specifically detects Clostridium Difficile Toxin A in Bacteria samples. It is validated for ELISA, Immunofluorescence
Specifications

Specifications

Clostridium Difficile Toxin A
0.2 mg/ml
ELISA, Immunofluorescence
Purified
RUO
10 mM PBS with 0.05% BSA
C. difficile Toxin A
Monoclonal
Protein A or G purified
253/422
ELISA 1-5 ug/ml, Immunocytochemistry/Immunofluorescence 1-2 ug/ml
Unconjugated
Mouse
Bacteria
tcdA, toxA
IgG1 Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit