Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Clostridium Perfringens Toxin Beta Sheep anti-Bacteria, Polyclonal, Novus Biologicals™
Sheep Polyclonal Antibody
Specifications
Antigen | Clostridium Perfringens Toxin Beta |
---|---|
Dilution | ELISA 1:100-1:2000 |
Applications | ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10062475
|
Novus Biologicals
NB10062475 |
0.25 mL | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
Clostridium Perfringens Toxin Beta Polyclonal specifically detects Clostridium Perfringens Toxin Beta in Bacteria samples. It is validated for ELISA.Specifications
Clostridium Perfringens Toxin Beta | |
ELISA | |
Unconjugated | |
Sheep | |
Bacteria | |
Culture filtrates of C. perfringens Type B/C, strains CN1240 and CN3424 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
ELISA 1:100-1:2000 | |
Polyclonal | |
Antisera | |
RUO | |
Whole antisera with 0.09% Sodium Azide | |
IgG | |
Unpurified | |
This product recognizes C. perfringens toxin beta, a peptide commonly associated with cases of food poisoning. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title