Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clostridium Tetani Toxoid Mouse anti-Bacteria, Alexa Fluor™ 405, Clone: 60000000, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP223554AF405

Catalog No. NP2234A40


Add to cart

Description

Description

Clostridium Tetani Toxoid Monoclonal antibody specifically detects Clostridium Tetani Toxoid in Bacteria samples. It is validated for ELISA.
Specifications

Specifications

Clostridium Tetani Toxoid
60000000
ELISA
Clostridium tetani toxoid, Tentoxylysin, Tetanus toxin, tetX
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Bacteria
ELISA
Alexa Fluor™ 405
50 mM sodium borate with 0.05% sodium azide
TETX
Tetanus toxoid adsorbed to aluminum hydroxide gel.
0.1 mL
Primary
Clone 6E7 binds to both tetanus toxin and tetanus toxoid
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit