Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAZ/WWTR1 Antibody - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
$204.00 - $506.50
Specifications
Antigen | Clostridium Tetani Toxoid |
---|---|
Clone | 14F5 |
Concentration | 1.0 mg/mL |
Dilution | ELISA 1:8000 |
Applications | ELISA |
Description
Clostridium Tetani Toxoid Monoclonal specifically detects Clostridium Tetani Toxoid in Bacteria, C. tetani samples. It is validated for ELISA.Specifications
Clostridium Tetani Toxoid | |
1.0 mg/mL | |
ELISA | |
Unconjugated | |
Mouse | |
Bacteria, C. tetani | |
TETX | |
IgG1 κ | |
Protein A or G purified | |
Reacts specifically with Clostridium tetani toxoid and toxin in a neutralizing manner. |
14F5 | |
ELISA 1:8000 | |
Monoclonal | |
Purified | |
RUO | |
Clostridium tetani toxoid, Tentoxylysin, Tetanus toxin, tetX | |
Tetanus toxoid adsorbed to aluminum hydroxide gel. | |
Primary | |
Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title