Thermo Scientific™ Closures with Prefitted Septa for 13mm Crimp/Snap-Ring LoVials™ Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ National Closures with Prefitted Septa are available in easy-to-apply Kim-Snap or standard aluminum variety to suit your application needs.
Manufacturer: Thermo Scientific™ C40154A
 View more versions of this product
Catalog No. 03-397-33
  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Septum Red PTFE/White Silicone
    Vial Closure Size 13mm
    Type Crimp Cap
    Item Description C4015-4A
    Thermo Scientific™ National Closures with Prefitted Septa are available in easy-to-apply Kim-Snap or standard aluminum variety to suit your application needs.
    • Kim-Snap™ closures provide a tight seal without the need of a crimper
    • National Scientific aluminum crimp closures provide a secure, leak-resistant seal
    • Aluminum seals must be applied with a crimping tool
  • Description & Specifications
Related Products
Product Content Feedback