Learn More
Thermo Scientific™ Closures with Prefitted Septa for 13mm Crimp/Snap-Ring LoVials™
Thermo Scientific™ National Closures with Prefitted Septa are available in easy-to-apply Kim-Snap or standard aluminum variety to suit your application needs.
Manufacturer:  Thermo Scientific™ C401553
 View more versions of this product
Catalog No. 03-397-36
  Check Availability
Add to cart
Description
Description
Thermo Scientific™ National Closures with Prefitted Septa are available in easy-to-apply Kim-Snap or standard aluminum variety to suit your application needs.
  • Kim-Snap™ closures provide a tight seal without the need of a crimper
  • National Scientific aluminum crimp closures provide a secure, leak-resistant seal
  • Aluminum seals must be applied with a crimping tool
Specifications
Specifications
Red PTFE/White Silicone/Red PTFE
13mm
Snap Cap
C4015-53