Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clotrimazole, Powder, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
23593-75-1
C22H17ClN2
344.84
VNFPBHJOKIVQEB-UHFFFAOYSA-N
1-[(2-chlorophenyl)diphenylmethyl]-1H-imidazole
ClC1=CC=CC=C1C(N1C=CN=C1)(C1=CC=CC=C1)C1=CC=CC=C1
23593-75-1
C22H17ClN2
344.84
VNFPBHJOKIVQEB-UHFFFAOYSA-N
1-[(2-chlorophenyl)diphenylmethyl]-1H-imidazole
ClC1=CC=CC=C1C(N1C=CN=C1)(C1=CC=CC=C1)C1=CC=CC=C1

CL127, 23593-75-1, C22H17ClN2

Spectrum™ Clotrimazole, Powder, USP is an azole antifungal used to treat skin infections. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Ignition Residue 0.1%
Loss on Drying 0.5%
Quantity 100 g
Packaging Amber Glass Bottle
CAS Max % 100%
Percent Purity 98-102%