Learn More
American Educational Products Cloud Formation Demonstration Apparatus
SCI_ED
Make clouds even on days when Mother Nature doesn't!
Manufacturer:  American Educational Products 7301
Catalog No. S41858
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Includes: Special flask, instructions
Description
Description
Bright blue cloudless skies are wonderful if you're going on a picnic or to a ball game. Unfortunately, they're not so great if you're trying to study cloud formation.
  • Special flask is fitted with a rubber hose, pinchcock and thick-walled rubber bulb
  • Rubber bulb holds water that nearly saturates air in flask
  • Smoke particles introduced form a nucleus of tiny water droplets
  • Bulb forms clouds when squeezed and released
Specifications
Specifications
Special flask, instructions, Glass flask connected to Rubber hose, pinch clamp, thick walled Rubber bulb
0.45 kg
Cloud Apparatus
1 lb.