Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clove Oil, FCC, 85%, Spectrum™

Click to view available options
Quantity:
25 mL
125 mL
500 mL
15 kg
Packaging:
Amber Glass Bottle
Steel Pail
8000-34-8
C7H12ClN3O2
205.64
VEEIFXWJNCAVEQ-UHFFFAOYNA-N
methyl 2-amino-3-(1H-imidazol-5-yl)propanoate hydrochloride
Cl.COC(=O)C(N)CC1=CN=CN1
8000-34-8
C7H12ClN3O2
205.64
VEEIFXWJNCAVEQ-UHFFFAOYNA-N
methyl 2-amino-3-(1H-imidazol-5-yl)propanoate hydrochloride
Cl.COC(=O)C(N)CC1=CN=CN1

CL130, 8000-34-8

Spectrum™ Clove Oil, FCC is a flavor and fragarant ingredient. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Refractive Index 1.527-1.535
Quantity 25 mL
CAS Max % 100%
Percent Purity 85%