Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris™ CLP 290
K+/Cl- cotransporter 2 (KCC2) activator; orally bioavailable prodrug of CLP 257 (Cat. No.5242)
$271.11 - $1127.78
Specifications
Molecular Formula | C19H21FN4O3S |
---|---|
Purity | 98% |
Target | KCC2 Activators |
Chemical Name or Material | (Z)-5-Fluoro-2-((4-oxo-2-(tetrahydropyridazin-1(2 H)-yl)thiazol-5(4 H)-ylidene)methyl)phenyl pyrrolidine-1-carboxylate |
CAS | 1181083-81-7 |
Description
K+/Cl- Cotransporter 2 (KCC2) activator. Activates spinal inhibitory interneurons, restores physiological neuronal activity in the spinal cord and induces functional recovery in a mouse model of spinal cord injury. Also decreases blood vasopressin and glucose levels in mice with Streptozotocin (Cat. No. 1621) induced diabetes mellitus. Orally bioavailable prodrug of CLP 257 (Cat. No.5242).Specifications
C19H21FN4O3S | |
KCC2 Activators | |
1181083-81-7 |
98% | |
(Z)-5-Fluoro-2-((4-oxo-2-(tetrahydropyridazin-1(2 H)-yl)thiazol-5(4 H)-ylidene)methyl)phenyl pyrrolidine-1-carboxylate |
Safety and Handling
Recommended Storage : Store at -20°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title