Learn More
Aviva Systems Biology Corp CLPB Antibody - middle region (ARP53791_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP53791_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CLPB. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CLPB. Alternate names: FLJ13152, HSP78, SKD3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61129