Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLPP Mouse anti-Human, Clone: OTI1C7, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA502060

Catalog No. 50-167-3147


Add to cart

Description

Description

The protein encoded by this gene belongs to the peptidase family S14 and hydrolyzes proteins into small peptides in the presence of ATP and magnesium. The protein is transported into mitochondrial matrix and is associated with the inner mitochondrial membrane.
Specifications

Specifications

CLPP
Monoclonal
0.36 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q16740
DFNB81, PRLTS3
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
8192
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI1C7
Unconjugated
CLPP
Liquid
CLPP
Full length human recombit protein of human CLPP produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit