Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLPP Mouse anti-Human, Rat, Canine, Monkey, Allophycocyanin/Cy7, Clone: OTI1F6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271363APCCY7

Catalog No. N271363APC7


Add to cart

Description

Description

CLPP Monoclonal antibody specifically detects CLPP in Human, Rat, Canine, Monkey samples. It is validated for Flow Cytometry.
Specifications

Specifications

CLPP
OTI1F6
Flow Cytometry
Affinity Purified, Conjugated
CLPP
Full length recombinant protein of human CLPP (NP_006003) produced in HEK293T cells.
Immunogen affinity purified
Core ESC Like Genes, Lipid and Metabolism, Stem Cell Markers
Primary
8192.0
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
ATP-dependent protease ClpAP, proteolytic subunit, human, ClpP (caseinolytic protease, ATP-dependent, proteolytic subunit, E. coli)homolog, ClpP caseinolytic peptidase, ATP-dependent, proteolytic subunit homolog (E.coli), ClpP caseinolytic protease, ATP-dependent, proteolytic subunit homolog, ClpP caseinolytic protease, ATP-dependent, proteolytic subunit homolog (E.coli), EC 3.4.21.92, Endopeptidase Clp, putative ATP-dependent Clp protease proteolytic subunit, mitochondrial
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Canine, Human, Monkey, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit