Learn More
CLPP Mouse anti-Canine, Human, Non-human primate, Rat, Clone: OTI1F6, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA525330
Catalog No. PIMA525330
Add to cart
Description
Description
The protein encoded by this gene belongs to the peptidase family S14 and hydrolyzes proteins into small peptides in the presence of ATP and magnesium. The protein is transported into mitochondrial matrix and is associated with the inner mitochondrial membrane.
Specifications
Specifications
CLPP
Monoclonal
0.81 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q16740
CLPP; ClpP caseinolytic protease, ATP-dependent, proteolytic subunit homolog (E. coli); ATP-dependent proteolytic subunit E.coli homolog; ATP-dependent protease ClpAP (E. coli), proteolytic subunit, human; ClpP (caseinolytic protease, ATP-dependent, proteolytic subunit, E. coli) homolog; ClpP caseinolytic peptidase, ATP-dependent, proteolytic subunit homolog (E. coli); ATP-dependent protease ClpAP, proteolytic subunit, human; ClpP caseinolytic protease, ATP-dependent, proteolytic subunit homolog
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
301117, 611738, 699593, 8192
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI1F6
Unconjugated
CLPP
Liquid
CLPP
Full length human recombit protein of CLPP produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human, Non-human primate, Rat