Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLPSL1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP257787

Catalog No. NBP257787


Add to cart

Description

Description

CLPSL1 Polyclonal antibody specifically detects CLPSL1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

CLPSL1
Unconjugated
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
C6orf127, Chromosome 6 Open Reading Frame 127, Colipase Like 1, Colipase-Like 1, Colipase-Like Protein 1, Colipase-Like Protein C6orf127, DJ510O8.6, ESP32
Rabbit
IgG
100 ul
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Immunohistochemistry (Paraffin)
Immunohistochemistry-Paraffin 1:200 - 1:500
Affinity Purified
CLPSL1
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:RGSLSPTKYNLLELKESCIRNQDCETGCCQRAPDNCESHCAEKGSEGSLCQ
Affinity purified
RUO
Primary
340204
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only