Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CLPTM1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546801
Description
Peptide sequence: ALTPTPAPTT TTATREEAST SLPTKPTQGA SSASEPQEAP PKPAEDKKKD.
CLPTM1 may play a role in T-cell development.
Specifications
CLPTM1 | |
Polyclonal | |
Unconjugated | |
CLPTM1 | |
cleft lip and palate associated transmembrane protein 1; cleft lip and palate transmembrane protein 1; Cleft lip and palate transmembrane protein 1 homolog; CLPTM1; CLPTM1, transmembrane protein; HS9; N14; Thymic epithelial cell surface antigen | |
Rabbit | |
Affinity Chromatography | |
RUO | |
1209 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O96005 | |
CLPTM1 | |
Synthetic peptide directed towards the C-terminal of human CLPTM1 (aa 620-669). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction