Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clr-a Rat anti-Mouse, Clone: 2000, MilliporeSigma™

Rat Monoclonal Antibody

$153.40 - $387.00

Specifications

Antigen Clr-a
Clone 2000
Concentration 0.5 mg/mL
Applications Immunohistochemistry, Western Blot
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABF2209100
View Documents
MilliporeSigma
MABF2209100UL
100 μL
Each for $387.00
Add to cart
 
MABF220925
View Documents
MilliporeSigma
MABF220925UL
25 μL
Each for $153.40
Add to cart
 
Description

Description

Clr-a Monoclonal antibody specifically detects Clr-a in Mouse samples. It is validated for Immunohistochemistry, Western Blotting.
Specifications

Specifications

Clr-a
0.5 mg/mL
Unconjugated
Rat
Mouse
c-Myc/His-tagged recombinant fragment corresponding to 140 amino acids from the extracellular domain of murine C-type lectin domain family 2 member E (Clr-a).
Monoclonal
Protein G purified
2000
Immunohistochemistry, Western Blot
Purified
RUO
Purified rat monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
IgG2aκ
Primary
Stable for 1 year at 2-8°C from date of receipt.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit