Learn More
CLRN2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 239941AP
Catalog No. 50-173-4195
Add to cart
Description
Description
Clarin 2 belongs to the clarin family of genes. It ft.s a small integral membrane glycoproteins with four transmembrane domains.
Specifications
Specifications
CLRN2
0.35 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
A0PK11
clarin 2, CLRN2
Rabbit
IgG
150 μL
-20°C
Primary
624224, 645104
Western Blot
Unconjugated
CLRN2
Liquid
Clrn2
CLRN2 Fusion Protein Ag21182
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse