Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CLSTN1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5140295
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: TITREVEPEG DGAEDPTVQE SLVSEEIVHD LDTCEVTVEG EELNHEQESL EVDMARLQQK GIEVSSSELG MTFTGVDTMA SYEEVLHLLR YRNWHARSLL DRKFKL.
CLSTN1 induces KLC1 association with vesicles and functions as a cargo in axonal anterograde transport. Complex formation with APBA2 and APP, CLSTN1 stabilizes APP metabolism and enhances APBA2-mediated suppression of beta-APP40 secretion, due to the retardation of intracellular APP maturation. In complex with APBA2 and C99, a C-terminal APP fragment, CLSTN1 abolishes C99 interaction with PSEN1 and thus APP C99 cleavage by gamma-secretase, most probably through stabilization of the direct interaction between APBA2 and APP. The intracellular fragment AlcICD suppresses APBB1-dependent transactivation stimulated by APP C-terminal intracellular fragment (AICD), most probably by competing with AICD for APBB1-binding.CLSTN1 may modulate calcium-mediated postsynaptic signals.
Specifications
CLSTN1 | |
Polyclonal | |
Unconjugated | |
CLSTN1 | |
1810034E21Rik; alcadein alpha; alcadein-alpha; alc-alpha; alcalpha1; alcalpha2; Alzheimer-related cadherin-like protein; cadherin-related family member 12; calsyntenin 1; calsyntenin-1; CDHR12; CLSTN1; clstn1 protein; CS1; CST-1; CSTN1; C-terminal fragment 1-alpha; CTF1-alpha; FLJ32258; fm67g03; Kiaa0911; Non-classical cadherin XB31alpha; PIK3CD; SAlc-alpha; Soluble Alc-alpha; wu:fk52g02; wu:fm67g03; XB31alpha; zgc:153744; zgc:56528 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22883 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O94985 | |
CLSTN1 | |
Recombinant protein corresponding to Human CLSTN1. Recombinant protein control fragment (Product #RP-109647). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction