Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLSTN2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549245

Catalog No. PA549245

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

May modulate calcium-mediated postsynaptic signals.
Specifications

Specifications

CLSTN2
Polyclonal
PBS with 0.09% sodium azide
Q9H4D0, Q9ER65
Calsyntenin-2, Alcadein-gamma, Alc-gamma, CLSTN2, CS2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
64084, 64085
Western Blot
Unconjugated
CLSTN2
Liquid
CLSTN2
KLH conjugated synthetic peptide between 743-770 amino acids from the Central region of human CLSTN2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit