Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clusterin/APOJ Mouse anti-Human, Janelia Fluor 646, Clone: 1A11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP104280JF646

Catalog No. NBP14280646

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Clusterin/APOJMonoclonalantibody specifically detects Clusterin/APOJ in Human samples.
Specifications

Specifications

Clusterin/APOJ
1A11
Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
CLU
Recombinant human Clusterin (23-449aa) purified from E. coli (NP_001822)
Protein G purified
Apoptosis, Cancer, Cardiovascular Biology, Cytokine Research, Dendritic Cell Markers, Hypoxia, Immunology, Innate Immunity
Primary
1191
Human
Immunohistochemistry, Immunohistochemistry (Paraffin)
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
40, 40, sulfated glycoprotein 2, Aging-associated gene 4 protein, aging-associated protein 4, apo-J, APOJ, Apolipoprotein J, CLIclusterin (complement lysis inhibitor, SP-40, clusterin, Complement cytolysis inhibitor, complement lysis inhibitor, Complement-associated protein SP-40, Ku70-binding protein 1, KUB1SGP2, MGC24903, NA1/NA2, SGP-2, SP-40, sulfated glycoprotein 2, Testosterone-repressed prostate message 2, testosterone-repressed prostate message 2, apolipoprotein J), TRPM-2TRPM2
Mouse
IgG1 Kappa
0.1mL
Store at 4°C in the dark.
Monoclonal
Anti-human Clusterin mAb, clone 1A11, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human Clusterin protein.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit