Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CMBL Mouse anti-Human, Alexa Fluor™ 750, Clone: 2B11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP222618AF750

Catalog No. NP222618A70


Add to cart

Description

Description

CMBL Monoclonal antibody specifically detects CMBL in Human samples. It is validated for Western Blot,ELISA.
Specifications

Specifications

CMBL
2B11
Western Blot, ELISA
carboxymethylenebutenolidase homolog, carboxymethylenebutenolidase homolog (Pseudomonas), carboxymethylenebutenolidase-like (Pseudomonas), EC 3.1, FLJ23617
Mouse
IgG2a Kappa
Store at 4°C in the dark.
Monoclonal
Human
ELISA, Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
CMBL
Recombinant human CMBL (1-245aa) purified from E. coli
0.1 mL
Primary
134147
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit