Learn More
CMC Rescue™ Anchor Plate
Manufacturer:  CMC Rescue™ 300615
 View more versions of this product
Catalog No. 19-808-468
Add to cart
Description
Description
  • Choose from aluminum or durable, corrosion-resistant stainless-steel construction
  • Prevents carabiners from jamming or clipping carabiner into wrong loop of anchor webbing
  • Serves as excellent collection point for anchor systems
  • Easier to see for faster, more accurate safety checks
Order Info
Hole Diameter: 0.88 in. inch (small); 2 in. (large). Weight: Stainless steel, 16 oz.; aluminum, 6 oz.
Certifications
UL listed, UL Classified NFPA 1983 - General Use.
Specifications
Specifications
1 lb.
Carabiner
Anchor Plate
0.45kg
Steel
Safety