Learn More
CMC Rescue™ 8 Descenders with Ears
Large enough to allow a safe lock off with larger diameter ropes
Manufacturer:  CMC Rescue™ 300850
Catalog No. 19-808-470
Add to cart
Description
Description
  • Carabiner hole roomy enough for extra carabiners
  • Nickel-chrome plated steel model performs exceptionally well in high-wear situations such as helicopter operations
Order Info
Diameter: 3 in. (large hole). Weight: steel, 20 oz.; aluminum, 6.6 oz.
Certifications
UL Classified to NFPA 1983, General Use.
Specifications
Specifications
Safety
Steel
Chrome
UL, NFPA 1983
3 oz.
Descender
5170 lbf.
3 in. dia. Hole, ears
13, 9.5 to 13mm rope
85g