Learn More
CMC Rescue™ Alta Flex™ Elbow Pads
Protect elbows while crawling in confined spaces, caves or other rescue areas
Manufacturer:  CMC Rescue™ 355310
Catalog No. 19-808-525
Add to cart
Description
Description
  • Nonslip rubber caps grip the surface, allowing faster movement
  • Two elastic straps with AltaGrip hook-and-loop fastening system ensure a proper fit
  • Easy-to-use buckle-fastening system allows for comfortable fit and minimizes interference with circulation
  • Color: Black
  • Weight: 190g (6.7 oz.)
Valuable for horizontal confined-space entries, cave rescue or tactical responses
Specifications
Specifications
Safety
Black
NFPA
190g
Elbow Pad
Two elastic straps, AltaGrip™ hook and loop fastening system
6.7 oz.