Learn More
CMC Rescue™ Edge Pads
Protect ropes from abrasion and help keep them clean, increasing their service life
Manufacturer:  CMC Rescue™ 294018
 View more versions of this product
Catalog No. 19-822-073
Add to cart
Description
Description
  • Reduce the amount a rope digs into a soft edge, such as the berm of a road
  • Made from rugged, heavy-duty, natural-color, 814g (24 oz.), No. 4 canvas
  • Corner grommets enable anchoring in place when needed
  • Grommets are offset, allowing for overlap when joining pads to extend coverage
Specifications
Specifications
Edge Pad
Large
Rope
86cm
71cm
680g
Canvas
Natural
34 in.
28 in.
24 oz.