Learn More
CMC Rescue™ Fire-Rescue Harness
With new fall arrest indicator and work-positioning side D-rings
Manufacturer:  CMC Rescue™ 202824
Catalog No. 19-808-406
Add to cart
Description
Description
  • Consists of multiple D-ring attachment points
  • JackBack™ removable padding with breathable D3 cloth keeps shoulder straps separated for easy donning
  • Dorsal D-ring allows adjustment and comfort for different size rescuers by staying in place when loaded
  • Front waist D-ring allows comfortable, stable sitting position for rappels
  • Sternum D-ring works well for helicopter hoists, tower climbing and limited fall protection
  • Gear loops for quick access to equipment
  • Quick-connect shoulder straps and leg loops make simple and fast donning of harness
  • Web-Keepers™ secures loose ends of the web
  • Pads on the leg loops can be adjusted
  • Front lift assembly accepts screw links for adding chest ascender or safety lanyards
  • Fall arrest indicator is located at the back of the harness
  • Mostly used by fire department rescue teams, USAR teams, helicopter crews and wilderness SAR teams
Certifications
UL Listed to NFPA 1983 – Class III and ANSI™ Z359.1.
Specifications
Specifications
34 to 46 in.
Safety
4 lb.
86 to 117 cm
Large/X-Large
Use for fire rescue