Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CMC Rescue™ Multi-Loop Straps

Tie into your anchor with just the right length of web

Manufacturer:  CMC Rescue™ 201013

 View more versions of this product

Catalog No. 19-808-344


Add to cart

Description

Description

  • Made of 25mm (1 in.) mil-spec tubular web
  • Handy for clipping several carabiners into one anchor point
  • Overall breaking strength of 22.2kN (5000 lbf.) with pocket breaking strength of 5kN (1100 lbf.)

Specifications

Specifications

Safety
Medium
Yellow
Carabiner
91cm
85g
8 Loop Strap
5000 lbf.
NFPA
36 in.
3 oz.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit