Learn More
CMC Rescue™ New England KM III Static Kernmantle Rescue Ropes
Torque-balanced to eliminate any spin during a rappel
Manufacturer:  CMC RESCUE K04160
 View more versions of this product
Catalog No. 19-822-087
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Polyester, Nylon
9900 lbf.
2400 in.
0.5 in.
7 lb.
Black
NFPA 1983 ,CE,EN 1891
6096cm
1.3cm
3.17kg