Learn More
CMC Rescue™ Pro Series™ Anchor Straps
Two layers of durable mil-spec nylon web help to resist tears and abrasion
Manufacturer:  CMC Rescue™ 201026
Catalog No. 19-808-351
Add to cart
Alert:
Use edge protection when appropriate, and observe safe carabiner-loading practices to maintain system strength.
Description
Description
  • Feature forged steel D-rings individually proof loaded and sized for easy choker (girth-hitch) configuration when desired
  • Offer breaking strength of 76kN (17,085 lbf.) in basket (U) configuration and 35kN (7868 lbf.) in end-to-end or choker configurations
  • 4.57m (15 ft.) 2X-Large straps come in blue
Certifications
UL Classified
Specifications
Specifications
179 in.
Mil-spec Nylon web
Safety
0.9kg
UL, NFPA 1983
457 cm
Anchor Strap
2 lb.
Blue