Learn More
CMC Rescue™ ProSeries™ Harnesses
Unique contoured padding pattern and breathable material on the waist and leg provide snug, comfortable fit while allowing more air circulation
Manufacturer:  CMC Rescue™ 202174
Catalog No. 19-808-377
Add to cart
Description
Description
  • Front lift design provides increased comfort by creating better weight distribution
  • D3 cloth construction for ultimate breathability and unparalleled comfort
  • Web-Keepers™ create secure place for web ends coming out of buckles
  • Molded padding for better fit, comfort and support
  • Large front D-Ring easily accepts three rescue carabiners
  • Quick-connect buckles on legs makes donning and doffing fast and easy
  • 1.75 in. web, used for adjusting straps, wraps completely around for ultimate strength
  • Unique comfortable padding construction protects stitch from abrasion
  • Black web with blue padding
  • Attachment points: front and rear waist
  • Designed for rope rescue and rappelling
Certifications
UL listed to NFPA 1983 - Class II
Specifications
Specifications
Safety
Black; Blue
Regular
Front
UL Classified to NFPA, Class II
30 to 44 in.
4 lb.