Learn More
CMC Rescue™ ProSeries™ Lifesaver Victim Harness
Attaches quickly and securely to victim no matter how or where they are
Manufacturer:  CMC Rescue™ 202405
Catalog No. 19-808-398
Add to cart
Includes: Wider 2 in. hook-and-loop Web Keeper™ to secure web before deployment.
Description
Description
  • Colored web makes usage of harness easier
  • Original V-ring and snap design allow harness to be put on without victim having to step into harness
  • For controlling victim's position, tie-in point get doubles and acts as “handle”
  • One size fits small children to large adults
  • Storage bag attached to harness prevents tangling when accessing victim
Specifications
Specifications
Safety
3 lb.