Learn More
CMC Rescue™ Pulleys
Manufacturer:  CMC Rescue™ 300323
 View more versions of this product
Catalog No. 19-808-462
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Large carabiner hole accepts multiple carabiners
  • Flat bottom manages Prusik hitches and works with Gibbs ascenders
  • Thicker sideplates with smooth rounded edges
  • Anodized aluminum wheels with Oilite™ bushings
Certifications
UL listed, UL classified to NFPA 1983, General Use.
Specifications
Specifications
Safety