Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CMC Rescue™ Rappel Gloves

Durable, double-layer gloves for tying knots, working with hardware or rappelling

$55.44

Specifications

Color Black, Tan
Length (Metric) 17.8 to 20.3cm
Material Leather, Spandex™
For Use With (Application) to tie knots and work with hardware, set up a system or to rappel
View More Specs

Products 5
Catalog Number Mfr. No. Circumference (Metric) Hand Length (English) Size Price Quantity & Availability  
Catalog Number Mfr. No. Circumference (Metric) Hand Length (English) Size Price Quantity & Availability  
19-100-325 View Documents CMC Rescue™
250203
22.8 to 25.4cm 9 to 10 in. Medium Pair for $55.44
Dozen of 12 PR for $1,205.45
Add to cart
 
19-100-326 View Documents CMC Rescue™
250204
25.4 to 27.9cm 10 to 11 in. Large Pair for $55.44
Add to cart
 
19-100-323 View Documents CMC Rescue™
250201
17.78 to 20.3cm 7 to 8 in. X-Small Pair for $55.44
Add to cart
 
250202 View Documents CMC Rescue™
250202
20.3 to 22.2cm 8 to 9 in. Small Pair for $55.44
Add to cart
 
19-100-327 View Documents CMC Rescue™
250205
27.9 to 30.4cm 11 to 12 in. X-Large Pair for $55.44
Add to cart
 
Description

Description

  • Pittard's Armor-Tan leather and unique Kevlar™ fiber stitching combine for durability
  • Double layer at thumb web protects hands when rappelling
  • Wear-resistant double palm guards against heat, moisture, cold and abrasion and eases grip fatigue
  • Elastic at wrist provides secure fit
  • When not in use, gloves can hang on harness gear loops using carabineer loops sewn into wrist
  • Space on label for user's name or initials
  • Color: Tan/Black
  • Length: 17.8 to 20.3cm (7 to 8 in.)

Specifications

Specifications

Black, Tan
17.8 to 20.3cm
Leather, Spandex™
to tie knots and work with hardware, set up a system or to rappel
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit