Learn More
CMC Rescue™ Rescue Harness
Meets need of search and rescue better than recreational climbing harness
Manufacturer:  CMC Rescue™ 202114
 View more versions of this product
Catalog No. 19-808-367
Add to cart
Description
Description
  • Even when hanging in harness, D-ring attachment point makes connecting carabiners easy
  • Extra wide waist and leg loops creates “work platform” than slim profile of climbing harness
  • Waist and leg pads with breathable D3 cloth for greater comfort
  • Rear waist D-ring for travel restraint and gear loops on each side
  • For rope rescue and rappelling
Certifications
UL listed to NFPA 1983 - Class II
Specifications
Specifications
Safety
32 to 42 in.
3 lb.
UL Classified to NFPA Standard 1983
Regular
Rear Waist