Learn More
CMC Rescue™ Riggers Gloves
SDP
A synthetic leather palm combines the dexterity of a lighter glove, with the protection of a heavier leather glove. The pre-curved, anatomic design ensures a close and comfortable fit for working with knots or hardware.
Manufacturer:  CMC Rescue™ 250305
 View more versions of this product
Catalog No. 19-100-337
Add to cart
Description
Description
  • Synthetic leather palm ensures dexterity and a high level of protection
  • Pre-curved, anatomic design offers a close and comfortable fit
  • Mesh back adds comfort, while also helping to keep the hands cool
  • Elastic wrist provides a secure fit
  • Carabiner loops allow for convenient clipping onto harness gear when not in use
Specifications
Specifications
Mesh
Black
Synthetic Leather
X-Large
Elasticated
Uncoated
Specific
Extrication Gloves
Belay Systems, Power tool Operation, Main Line Hauling, and Repelling
25030